4-(Carboxymethyl)phenylboronsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

444670010
ACRO444670010EA 145 EUR
ACRO444670010
4-(Carboxymethyl)phenylboronsäurepinakolester 97%
4-(Carboxymethyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₄
Schmelzpunkt: 166…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038755
CAS-Nummer: 797755-07-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 162 to 168 °C
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR