4-Chlorphenylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(4-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chlorophenylboronic acid Pinacol ester, 4-Chlorobenzeneboronic acid pinacol ester, 1-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
444690050
444690250
ACRO444690050EA
64.9
EUR
ACRO444690050
ACRO444690250
4-Chlorphenylboronsäurepinakolester 97%
4-Chlorphenylboronsäurepinakolester
Formel:
C₁₂H₁₆BClO₂ Schmelzpunkt: 55…56 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663875 CAS-Nummer: 195062-61-4 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid or crystalline solid |
Infrared spectrum | Conforms |
Melting point | 54 to 59 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...