2-Chloropyrimidine-5-boronic acid pinacol ester 95%

Lieferant: Thermo Fisher Scientific
Achtung

444700010
ACRO444700010EA 104 EUR
ACRO444700010
2-Chloropyrimidine-5-boronic acid pinacol ester 95%
2-Chloropyrimidine-5-boronic acid pinacol ester
Formel: C₁₀H₁₄BClN₂O₂
Schmelzpunkt: 67…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11856048
CAS-Nummer: 1003845-08-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to orange Solid or crystalline solid
Infrared spectrum Conforms
Melting point 63 to 69 °C
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR