3-Iod-1H-pyrazol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Iodopyrazole

444810050
ACRO444810050EA 338 EUR
ACRO444810050
3-Iod-1H-pyrazol 97%
3-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193.98 g/mol
Schmelzpunkt: 68…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12022324
CAS-Nummer: 4522-35-4

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 67 to 74 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR