4-Carbamoylphenylboronic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

444900050 444900250
ACRO444900050EA 141 EUR
ACRO444900050 ACRO444900250
4-Carbamoylphenylboronic acid 97%
4-Carbamoylphenylboronic acid
Formel: C₇H₈BNO₃
Schmelzpunkt: 242…252 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411940
CAS-Nummer: 123088-59-5

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR