4-(N,N-Dimethylamino)phenylboronsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

444960010 444960050
ACRO444960010EA 76.1 EUR
ACRO444960010 ACRO444960050
4-(N,N-Dimethylamino)phenylboronsäurepinakolester 97%
4-(N,N-Dimethylamino)phenylboronsäurepinakolester
Formel: C₁₄H₂₂BNO₂
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663854
CAS-Nummer: 171364-78-6

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light green to tan Powder or solid
Infrared spectrum Conforms
Melting point 122 to 127 °C
GC ≥96 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR