6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N,N-Dimethyl-3-((5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)oxy)propan-1-amine, 2-(3-N,N-Dimethylamino-propoxy)pyridine-5-boronic acid, pinacol ester, DIMETHY(3-[5-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)PYRIDIN-22-YLOXY]PROPYL)AMINE

444970050
ACRO444970050EA 1397 EUR
ACRO444970050
6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester 97%
6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester
Formel: C₁₆H₂₇BN₂O₃
Schmelzpunkt: 34…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797989
CAS-Nummer: 918643-56-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 33 to 39 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR