4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 4,4-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-enone
444980010
ACRO444980010EA
354
EUR
ACRO444980010
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester 97%
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester
Formel:
C₁₄H₂₃BO₃ Schmelzpunkt: 99…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027074 CAS-Nummer: 219489-09-5 |
Spezifikation Testergebnisse
Appearance | White to yellow Solid |
Infrared spectrum | Conforms |
Melting point | 98 to 102 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...