4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid ethyl ester, Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, 2-[(4-Ethoxycarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Ethoxycarbonylmethyl)phenylboronic acid Pinacol ester

445000050
ACRO445000050EA 570 EUR
ACRO445000050
4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester 97%
4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
Formel: C₁₆H₂₃BO₄
Molecular Weight: 290,1624 g/mol
Schmelzpunkt: 41…43 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08704684
CAS-Nummer: 859169-20-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Low melting solid
Infrared spectrum Conforms
Melting point 38 to 46 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR