3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 2-hydroxy-4-(4,4,5,5-tetraMethyl-1,3,2-dioxaborolan-2-yl)benzoate

445050010
ACRO445050010EA 237 EUR
ACRO445050010
3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 97%
3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester
Formel: C₁₄H₁₉BO₅
Molecular Weight: 278,11 g/mol
Schmelzpunkt: 85…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458200
CAS-Nummer: 1073371-99-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Solid
Infrared spectrum Conforms
Melting point 84 to 90 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR