3-Hydroxy-4-methoxyphenylboronic acid pinacol ester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
445060010
ACRO445060010EA
264
EUR
ACRO445060010
3-Hydroxy-4-methoxyphenylboronic acid pinacol ester 97%
3-Hydroxy-4-methoxyphenylboronic acid pinacol ester
Formel:
C₁₃H₁₉BO₄ Schmelzpunkt: 101…102 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09027077 CAS-Nummer: 269410-23-3 |
Spezifikation Testergebnisse
Appearance | White to tan Solid |
Infrared spectrum | Conforms |
Melting point | 99 to 104 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...