4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific

445080010
ACRO445080010EA 278 EUR
ACRO445080010
4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester 97%
4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Solid
Infrared spectrum Conforms
Melting point 136 to 142 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR