1-Methyl-1H-indazol-4-boronsäurepinakolester 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
445130010
ACRO445130010EA
153
EUR
ACRO445130010
1-Methyl-1H-indazol-4-boronsäurepinakolester 97%
1-Methyl-1H-indazol-4-boronsäurepinakolester
Formel:
C₁₄H₁₉BN₂O₂ Molecular Weight: 258,1239 g/mol Schmelzpunkt: 83…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10700158 CAS-Nummer: 885698-94-2 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Melting point | 82 to 87 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...