Diclofenac-Natrium 98%

Lieferant: Thermo Fisher Scientific
Gefahr

445250100
ACRO445250100EA 91.2 EUR
ACRO445250100
Diclofenac-Natrium 98%
Formel: C₁₄H₁₀Cl₂NNaO₂
Schmelzpunkt: 283…285 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082251
CAS-Nummer: 15307-79-6
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %
pH 6.5 to 7.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR