3-Amino-4-fluorophenylboronic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Borono-2-fluoroaniline, Boronic acid, B-(3-amino-4-fluorophenyl)-, 3-Amino-4-fluorobenzeneboronic acid

445290010
ACRO445290010EA 269 EUR
ACRO445290010
3-Amino-4-fluorophenylboronic acid 97%
3-Amino-4-fluorophenylboronic acid
Formel: C₆H₇BFNO₂
Molecular Weight: 154.94 g/mol
Schmelzpunkt: 226…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03092925
CAS-Nummer: 873566-75-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR