2-Aminophenylboronsäure Hydrochlorid 97%

Lieferant: Thermo Fisher Scientific
Achtung

445310010 445310050
ACRO445310010EA 96.1 EUR
ACRO445310010 ACRO445310050
2-Aminophenylboronsäure Hydrochlorid 97%
2-Aminophenylboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
Molecular Weight: 173.4052 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258096
CAS-Nummer: 863753-30-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR