2-(Brommethyl)phenylboronsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-(Brommethyl)phenyl)metaborsäure, Dihydroxy(2-(brommethyl)phenyl)boran, 2-(Brommethyl)benzolboronsäure

445330050
ACRO445330050EA 247 EUR
ACRO445330050
2-(Brommethyl)phenylboronsäure 95%
2-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
Molecular Weight: 214,85 g/mol
Schmelzpunkt: 135…136,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318941
CAS-Nummer: 91983-14-1

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to off-white Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR