4-Cyan-2-fluorphenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Cyano-2-fluorophenylboronic Acid, 4-Cyano-2-fluorobenzeneboronic acid

445390010
ACRO445390010EA 364 EUR
ACRO445390010
4-Cyan-2-fluorphenylboronsäure 97%
4-Cyan-2-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164,93 g/mol
Schmelzpunkt: 304…304,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07644472
CAS-Nummer: 1150114-77-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR