4-(Cyanmethoxy)phenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-(Cyanomethoxy)phenyl)boronic acid

445400010
ACRO445400010EA 247 EUR
ACRO445400010
4-(Cyanmethoxy)phenylboronsäure 97%
4-(Cyanmethoxy)phenylboronsäure
Formel: C₈H₈BNO₃
Molecular Weight: 176,9700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09908170
CAS-Nummer: 947533-23-5

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR