4-(Ethoxycarbonylmethyl)phenylboronsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-[(Ethoxycarbonyl)methyl]phenylboronic acid, (4-(2-Ethoxy-2-oxoethyl)phenyl)boronic acid

445430010
ACRO445430010EA 437 EUR
ACRO445430010
4-(Ethoxycarbonylmethyl)phenylboronsäure 97%
4-(Ethoxycarbonylmethyl)phenylboronsäure
Formel: C₁₀H₁₃BO₄
Molecular Weight: 208.200 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08704683
CAS-Nummer: 92243-74-8

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to tan Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR