4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Fluoro-3-(1H-tetrazol-5-yl)phenyl)boronic acid, [4-Fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid, Boronic acid, B-[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]-
445460010
ACRO445460010EA
201
EUR
ACRO445460010
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid 95%
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid
Formel:
C₇H₆BFN₄O₂ Molecular Weight: 207,96 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13176533 CAS-Nummer: 1009303-56-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Additional info | May contain varying amounts of anhydrides |
HPLC | ≥94.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...