1-Methyl-1H-pyrazole-4-boronic acid hydrochloride 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Methylpyrazole-4-boronic acid, HCl, (1-Methyl-1H-pyrazol-4-yl)boronic acid hydrochloride

445490010
ACRO445490010EA 163 EUR
ACRO445490010
1-Methyl-1H-pyrazole-4-boronic acid hydrochloride 95%
1-Methyl-1H-pyrazole-4-boronic acid hydrochloride
Formel: C₄H₈BClN₂O₂
Molecular Weight: 162,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09972098
CAS-Nummer: 1026796-02-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white to light yellow Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR