6-Chinoxalincarbonsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Quinoxaline-6-boronic acid, pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline

445520010
ACRO445520010EA 180 EUR
ACRO445520010
6-Chinoxalincarbonsäurepinakolester 97%
6-Chinoxalincarbonsäurepinakolester
Formel: C₁₄H₁₇BN₂O₂
Molecular Weight: 256.1080 g/mol
Siedepunkt: 130…145 °C (.6 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD11054040
CAS-Nummer: 1167418-13-4

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Viscous liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR