2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,2,6,6-Tetramethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-3,6-dihydro-2H-pyran, (2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-yl)boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-(2,2,6,6-tetramethyl-3,6-dihydro-2H-pyran-4-yl)-1,3,2-dioxaborolane
445530010
ACRO445530010EA
366
EUR
ACRO445530010
2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester 97%
2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester
Formel:
C₁₅H₂₇BO₃ Molecular Weight: 266,19 g/mol Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044439 CAS-Nummer: 1142363-56-1 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Melting point | 91 to 96 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...