1-[(2-Thienyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-(Thiophen-2-ylmethyl)pyrazole-4-boronic acid, pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(2-thienylmethyl)-1H-pyrazole, 1H-Pyrazole,4-(4,4,5,5-tetramethyl-1,3,2-dioxaboro

445580010
ACRO445580010EA 583 EUR
ACRO445580010
1-[(2-Thienyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester 97%
1-[(2-Thienyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₄H₁₉BN₂O₂S
Molecular Weight: 290,19 g/mol
Schmelzpunkt: 57…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08706027
CAS-Nummer: 864754-19-8

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to yellow Solid
Infrared spectrum Conforms
Melting point 54 to 59 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR