Brom 99.5+% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

445871000 445875000
ACRO445871000EA 59.3 EUR
ACRO445871000 ACRO445875000
Brom 99.5+% ACS
Bromine
Formel: Br₂
Siedepunkt: 58,7 °C (760 mmHg)
Dichte: 3,111 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00010896
CAS-Nummer: 7726-95-6
UN: 1744
ADR: 8,I

Bestellen

Spezifikation Testergebnisse

Appearance Dark red-brown Liquid
Titration Iodometric ≥99.5 %
Residue after evaporation ≤0.005 %
Chlorine (Cl) ≤0.05 %
Iodine (I) ≤0.001 %
Nickel (Ni) ≤5 ppm
Heavy metals (as Pb) ≤2 ppm
Sulfur compounds (as S) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR