DL-Arginin-hydrochlorid, wasserfrei 98%

Lieferant: Thermo Scientific

Synonyme: DL-Arginine xhydrochloride, DL-Arginine Hydrochloride, DL-(+/-)-Arginine hydrochloride, DL-(±)-Arginine Hydrochloride, H-DL-Arg-OH.HCl, H-DL-Arg-OH·HCl, H-DL-(±)-Arg-OH·HCl, DL-2-Amino-5-guanidinopentanoic acid hydrochloride monohydrate

446020050
ACRO446020050EA 65.4 EUR
ACRO446020050
DL-Arginin-hydrochlorid, wasserfrei 98%
DL-Arginine hydrochloride
Formel: C₆H₁₄N₄O₂.HCl
Molecular Weight: 210,66 g/mol
Schmelzpunkt: 200…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064549
CAS-Nummer: 32042-43-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Assay 97.5 to 102.5 %
Loss on drying ≤3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR