Kaliumhydrogencarbonat ACS

Lieferant: Thermo Fisher Scientific

Synonyme: Kaliumbicarbonat

446300025 446301000 446305000
ACRO446300025EA 188 EUR
ACRO446300025 ACRO446301000 ACRO446305000
Kaliumhydrogencarbonat ACS
Kaliumhydrogencarbonat
Formel: KHCO₃
Molecular Weight: 100.12 g/mol
Schmelzpunkt: 292 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011402
CAS-Nummer: 298-14-6
Merck Index: 13,07691

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white Crystals or crystalline powder
Titration with HCl 99.7 to 100.5 % (On dried basis)
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤5 ppm
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.03 %
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR