2-Chloro-5-(trifluoromethyl)pyrazine 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
446480010
446480050
ACRO446480010EA
129
EUR
ACRO446480010
ACRO446480050
2-Chloro-5-(trifluoromethyl)pyrazine 97%
2-Chloro-5-(trifluoromethyl)pyrazine
Formel:
C₅H₂ClF₃N₂ Siedepunkt: 88 °C (118 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189478 CAS-Nummer: 799557-87-2 UN: 2922 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow to orange to brown Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...