4-(Trifluoromethyl)-1H-pyrazole 97%

Lieferant: Thermo Fisher Scientific
Achtung

446490010
ACRO446490010EA 379 EUR
ACRO446490010
4-(Trifluoromethyl)-1H-pyrazole 97%
4-(Trifluoromethyl)-1H-pyrazole
Formel: C₄H₃F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11226572
CAS-Nummer: 52222-73-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline solid
Infrared spectrum Conforms
Water ≤2.5 %
GC ≥96 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR