5-Bromo-3-fluoro-2-pyridine carboxylic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromo-3-fluoropicolinic acid, 5-Bromo-3-fluoropyridine-2-carboxylic acid

446530050
ACRO446530050EA 288 EUR
ACRO446530050
5-Bromo-3-fluoro-2-pyridine carboxylic acid 97%
5-Bromo-3-fluoro-2-pyridine carboxylic acid
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
Schmelzpunkt: 165…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07437865
CAS-Nummer: 669066-91-5

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to brown Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure
Titration with NaOH ≥96.0 to ≤104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR