2-(Trifluoromethyl)-1H-indole 97%
Lieferant: Thermo Scientific
|
Achtung
|
446560010
ACRO446560010EA
188
EUR
ACRO446560010
2-(Trifluoromethyl)-1H-indole 97%
2-(Trifluoromethyl)-1H-indole
Formel:
C₉H₆F₃N Schmelzpunkt: 107…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972104 CAS-Nummer: 51310-54-4 |
Spezifikation Testergebnisse
Appearance | White to yellow to light brown Powder or solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...