2,3-Difluoro-4-(trifluoromethyl)pyridine 97%

Lieferant: Thermo Scientific
Achtung

446780010
ACRO446780010EA 133 EUR
ACRO446780010
2,3-Difluoro-4-(trifluoromethyl)pyridine 97%
2,3-Difluoro-4-(trifluoromethyl)pyridine
Formel: C₆H₂F₅N
Siedepunkt: 82…84 °C (400Torr)
Lagertemperatur: Raumtemperatur
MDL: MFCD16610846
CAS-Nummer: 1215034-77-7
UN: 1993
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR