2',7'-Dichlorfluorescein ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
446930010
446930100
ACRO446930010EA
48.3
EUR
ACRO446930010
ACRO446930100
2',7'-Dichlorfluorescein ACS
2',7'-Dichlorfluorescein
Formel:
C₂₀H₁₀Cl₂O₅ Molecular Weight: 401.20 g/mol Schmelzpunkt: 280 °C (Zers.) Dichte: 0.79 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005047 CAS-Nummer: 76-54-0 |
Spezifikation Testergebnisse
Appearance | Light orange to red-brown Powder |
Suitability | (as adsorption indicator) Passes test |
Clarity of alcohol solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...