Kaliumbromat ACS

Lieferant: Thermo Fisher Scientific
Gefahr

446941000 446945000
ACRO446941000EA 38.5 EUR
ACRO446941000 ACRO446945000
Kaliumbromat ACS
Potassium bromate
Formel: KBrO₃
Schmelzpunkt: 350 °C
Dichte: 3,27 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011359
CAS-Nummer: 7758-01-2
UN: 1484
ADR: 5.1,II

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Titration Iodometric ≥99.8 % (Dried basis)
pH 5.0 to 9.0 (5% soln. at 25°C)
Bromide (Br) Passes test (Limit about 0.05%)
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.002 %
Sodium (Na) ≤0.01 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR