Kaliumbromat ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
446941000
446945000
ACRO446941000EA
38.5
EUR
ACRO446941000
ACRO446945000
Kaliumbromat ACS
Potassium bromate
Formel:
KBrO₃ Schmelzpunkt: 350 °C Dichte: 3,27 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011359 CAS-Nummer: 7758-01-2 UN: 1484 ADR: 5.1,II |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Titration Iodometric | ≥99.8 % (Dried basis) |
pH | 5.0 to 9.0 (5% soln. at 25°C) |
Bromide (Br) | Passes test (Limit about 0.05%) |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤0.002 % |
Sodium (Na) | ≤0.01 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...