D-(+)-Glucose-6-phosphate sodium salt 98%

Lieferant: Thermo Scientific

Synonyme: ROBISON ESTER MONOSODIUM SALT, Sodium (2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl hydrogenphosphate, G-6-P Na, BETA-D-GLUCOSE 6-PHOSPHATE MONOSODIUM SALT, D-Glucose 6-phosphate sodium

446980010
ACRO446980010EA 97.4 EUR
ACRO446980010
D-(+)-Glucose-6-phosphate sodium salt 98%
D-(+)-Glucose-6-phosphate sodium salt
Formel: C₆H₁₂NaO₉P
Molecular Weight: 282,12 g/mol
Schmelzpunkt: 192…197 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006611
CAS-Nummer: 54010-71-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤6 %
Specific optical rotation +31° to +35° (20°C, 589 nm) (c=10 in H2O)
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR