1-Chlorbutan 99.5% zur Analyse
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Butylchlorid
447020010
ACRO447020010EA
74.4
EUR
ACRO447020010
1-Chlorbutan 99.5% zur Analyse
1-Chlorbutan
Formel:
CH₃(CH₂)₃Cl Molecular Weight: 92.57 g/mol Siedepunkt: 77…78 °C (760 mmHg) Schmelzpunkt: –123 °C Dichte: 0.88 g/cm³ Flammpunkt: –12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001009 CAS-Nummer: 109-69-3 UN: 1127 ADR: 3 II Merck Index: 13,01559 |
|
Spezifikation Testergebnisse
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4010 to 1.4030 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥99.4 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Acidity | ≤0.0005 meq/g |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Sulfur compounds (as S) | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...