Ammoniumsulfat 99% für die Biochemie

Lieferant: Thermo Fisher Scientific

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

447130025 447135000
ACRO447130025EA 189 EUR
ACRO447130025 ACRO447135000
Ammoniumsulfat 99% für die Biochemie
Ammoniumsulfat
Formel: (NH₄)₂SO₄
Schmelzpunkt: 280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
UV (0.1 M in water)
pH 4.5 to 5.5 (5 % in water)
Titration excess NaOH ≥98.5 %
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Arsenic (As) ≤1 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤2 ppm
Zinc (Zn) ≤1 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR