4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde

447430050
ACRO447430050EA 552 EUR
ACRO447430050
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde
Formel: C₅H₅BrN₂O
Molecular Weight: 189,02 g/mol
Schmelzpunkt: 71…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00103235
CAS-Nummer: 287917-96-8

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR