Xylenolorange Tetranatriumsalz ACS
Lieferant: Thermo Fisher Scientific
447600010
447600050
447600250
ACRO447600010EA
44.6
EUR
ACRO447600010
ACRO447600050
ACRO447600250
Xylenolorange Tetranatriumsalz ACS
Xylenolorange Tetranatriumsalz
Formel:
C₃₁H₂₈N₂Na₄O₁₃S Schmelzpunkt: 210 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044293 CAS-Nummer: 3618-43-7 |
Spezifikation Testergebnisse
Appearance | Red-brown to dark brown Crystalline powder |
Suitability | (for Zn titration) Passes test |
Clarity of solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...