Natriumdihydrogenphosphat Monohydrat 98%, reinst
Lieferant: Thermo Fisher Scientific
Synonyme:
Natriumphosphat monobasisch Monohydrat
447740010
447740050
ACRO447740010EA
71.9
EUR
ACRO447740010
ACRO447740050
Natriumdihydrogenphosphat Monohydrat 98%, reinst
Natriumdihydrogenphosphat Monohydrat
Formel:
NaH₂PO₄·H₂O Schmelzpunkt: 100 °C Dichte: 2,04 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149208 CAS-Nummer: 10049-21-5 |
|
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals |
Loss on drying | 10.0 % to 15.0 % |
pH | 4.1 to 4.5 |
Titration with NaOH | ≥97.5 % |
Chloride (Cl) | ≤140 ppm |
Arsenic (As) | ≤8 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Insoluble matter | ≤0.2 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...