Natriumdihydrogenphosphat Monohydrat 98%, reinst

Lieferant: Thermo Fisher Scientific

Synonyme: Natriumphosphat monobasisch Monohydrat

447740010 447740050
ACRO447740010EA 71.9 EUR
ACRO447740010 ACRO447740050
Natriumdihydrogenphosphat Monohydrat 98%, reinst
Natriumdihydrogenphosphat Monohydrat
Formel: NaH₂PO₄·H₂O
Schmelzpunkt: 100 °C
Dichte: 2,04 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149208
CAS-Nummer: 10049-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Loss on drying 10.0 % to 15.0 %
pH 4.1 to 4.5
Titration with NaOH ≥97.5 %
Chloride (Cl) ≤140 ppm
Arsenic (As) ≤8 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.2 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR