Calciumacetat xHydrat 99% zur Analyse

Lieferant: Thermo Scientific

Synonyme: Calciumdi(acetat) Hydrat, Essigsäure Calciumsalz Hydrat, Calciumacetat Hydrat

447860010
ACRO447860010EA 54.8 EUR
ACRO447860010
Calciumacetat xHydrat 99% zur Analyse
Calciumacetat xHydrat
Formel: (H₃CCOO)₂Ca·nH₂O
Molecular Weight: 176,18 g/mol
Dichte: 1,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150019
CAS-Nummer: 114460-21-8

Bestellen

Spezifikation Testergebnisse

Appearance White Powder or granules
Infrared spectrum Conforms
Titration Complexometric ≥98.5 % (on dried substance)
Water ≤10 %
pH 6.0 to 9.0 (10 % in water)
Chloride (Cl) ≤0.05 %
Sulfate (SO4) ≤0.1 %
Arsenic (As) ≤3 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤2 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR