Natriumthiosulfat Pentahydrat 99+% Biochemistry

Lieferant: Thermo Fisher Scientific

Synonyme: Fixiersalz

447940010 447942500
ACRO447940010EA 133 EUR
ACRO447940010 ACRO447942500
Natriumthiosulfat Pentahydrat 99+% Biochemistry
Natriumthiosulfat Pentahydrat
Formel: Na₂S₂O₃.5H₂O
Molecular Weight: 248.19 g/mol
Schmelzpunkt: 48.5 °C
Dichte: 1.72 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00149186
CAS-Nummer: 10102-17-7
Merck Index: 12,08844

Bestellen

Spezifikation Testergebnisse

Appearance White Powder or crystals
Titration Iodimetric 99.0 to 101.0 %
Solubility (1 M in water) Clear colorless
pH 6.0 to 7.5 (5 % in water)
Chloride (Cl) ≤0.008 %
SO4 and SO3 (as SO4) ≤0.1 %
Calcium (Ca) ≤0.002 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR