Ammoniumiodid 99%, reinst

Lieferant: Thermo Fisher Scientific
Achtung

448070025 448071000 448075000
ACRO448070025EA 752 EUR
ACRO448070025 ACRO448071000 ACRO448075000
Ammoniumiodid 99%, reinst
Ammoniumiodid
Formel: NH₄I
Schmelzpunkt: 551 °C (Subl.)
Dichte: 2,51 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011424
CAS-Nummer: 12027-06-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder (May turn yellow on storage)
Titration Argentometric 98.5 to 101.5 %
Residue after ignition ≤0.05 %
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR