di-Natriumhydrogenphosphat Dihydrat ≥98%, reinst

Lieferant: Thermo Fisher Scientific

448150010 448150050
ACRO448150010EA 36.3 EUR
ACRO448150010 ACRO448150050
di-Natriumhydrogenphosphat Dihydrat ≥98%, reinst
di-Natriumhydrogenphosphat Dihydrat
Formel: Na₂HPO₄·2H₂O
Schmelzpunkt: 243…245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149182
CAS-Nummer: 10028-24-7

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white or colorless Crystalline powder or crystals
Loss on drying 19.5 % to 21.0 % (130°C)
Solubility (5 % in water) Clear colorless
Titration with HClO4 97.5 to 102.5 % (dried substance)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR