3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,3',5,5'-Tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione, 3,3',5,5'-Tetra-tert-butyldiphenoquinone
448300020
ACRO448300020EA
103
EUR
ACRO448300020
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone 98%
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone
Formel:
C₂₈H₄₀O₂ Molecular Weight: 408.63 g/mol Schmelzpunkt: 247…248 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051798 CAS-Nummer: 2455-14-3 |
Spezifikation Testergebnisse
Appearance | Yellow to brown to red Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...