Picolinamidin Hydrochlorid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Picolinimidamide hydrochloride, 2-Amidinopyridine hydrochloride

448610050
ACRO448610050EA 131 EUR
ACRO448610050
Picolinamidin Hydrochlorid 97%
Picolinamidin Hydrochlorid
Formel: C₆H₈ClN₃
Molecular Weight: 157,60 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052271
CAS-Nummer: 51285-26-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Solid
Infrared spectrum Conforms
Titration Argentometric ≥96.0 to ≤104.0 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR