5-Bromo-1,3,4-thiadiazol-2-amine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-5-bromo-1,3,4-thiadiazole, 1,3,4-Thiadiazol-2-amine, 5-bromo-, 5-bromo-1,3,4-thiadiazol-2-amine

448670100
ACRO448670100EA 337 EUR
ACRO448670100
5-Bromo-1,3,4-thiadiazol-2-amine 97%
5-Bromo-1,3,4-thiadiazol-2-amine
Formel: C₂H₂BrN₃S
Molecular Weight: 180,02 g/mol
Schmelzpunkt: 167…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00464325
CAS-Nummer: 37566-39-5
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 165 to 170 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR